1,5-Dimethyl-1H,5H-[1,2,4]triazolo[1,2-a][1,2,4]triazole-3,7-dithiol
Supplier: Apollo Scientific
OR23575-10G OR23575-1G OR23575-5G
APOSOR23575-10GEA 433 EUR
APOSOR23575-10G APOSOR23575-1G APOSOR23575-5G
1,5-Dimethyl-1H,5H-[1,2,4]triazolo[1,2-a][1,2,4]triazole-3,7-dithiol
1,5-Dimethyl-1H,5H-[1,2,4]triazolo[1,2-a][1,2,4]triazole-3,7-dithiol
Formula: C₆H₁₀N₄S₂
MW: 202.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00135059
CAS Number: 16085-50-0