1,3,5-Trimethyl-4-nitro-1H-pyrazole
Supplier: Apollo Scientific
Warning
Synonyms: 1,3,5-Trimethyl-4-nitro-1h-pyrazole
OR23875-1G OR23875-5G
APOSOR23875-1GEA 23.2 EUR
APOSOR23875-1G APOSOR23875-5G
1,3,5-Trimethyl-4-nitro-1H-pyrazole
1,3,5-Trimethyl-4-nitro-1H-pyrazole
Formula: C₆H₉N₃O₂
MW: 155.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00052533
CAS Number: 1125-30-0