1-(2-Aminoethyl)-2-methyl-5-phenyl-1H-pyrrole-3-carboxylic acid, 1-BOC protected
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
1-{2-[(tert-Butoxycarbonyl)amino]ethyl}-2-methyl-5-phenyl-1H-pyrrole-3-carboxylic acid, tert-Butyl [2-(3-carboxy-2-methyl-5-phenyl-1H-pyrrol-1-yl)ethyl]carbamate
OR2409-1G
OR2409-250MG
APOSOR2409-1GEA
434
EUR
APOSOR2409-1G
APOSOR2409-250MG
1-(2-Aminoethyl)-2-methyl-5-phenyl-1H-pyrrole-3-carboxylic acid, 1-BOC protected
1-(2-Aminoethyl)-2-methyl-5-phenyl-1H-pyrrole-3-carboxylic acid, 1-BOC protected
Formula:
C₁₉H₂₄N₂O₄ MW: 344.405 g/mol Melting Pt: 135…137 °C Storage Temperature: Ambient |
MDL Number:
MFCD01567245 CAS Number: 306936-27-6 UN: 3261 ADR: 8 III |