4-hydrazinothieno[2,3-d]pyrimidine
Supplier: Apollo Scientific
OR24397-1G OR24397-250MG OR24397-5G
APOSOR24397-1GEA 174 EUR
APOSOR24397-1G APOSOR24397-250MG APOSOR24397-5G
4-hydrazinothieno[2,3-d]pyrimidine
4-hydrazinothieno[2,3-d]pyrimidine
Formula: C₆H₆N₄S
Storage Temperature: Ambient
MDL Number: MFCD00728978
CAS Number: 14080-58-1