Diethyl 2-{[(1,3-dimethyl-1H-pyrazol-5-yl)amino]methylidene}malonate
Supplier: Apollo Scientific
OR25193-10G
OR25193-1G
OR25193-5G
APOSOR25193-10GEA
346
EUR
APOSOR25193-10G
APOSOR25193-1G
APOSOR25193-5G
Diethyl 2-{[(1,3-dimethyl-1H-pyrazol-5-yl)amino]methylidene}malonate
Diethyl 2-{[(1,3-dimethyl-1H-pyrazol-5-yl)amino]methylidene}malonate
Formula:
C₁₃H₁₉N₃O₄ MW: 281.308 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00067953 CAS Number: 20481-33-8 |