Ethyl 5-methyl-3-(methylthio)-4,4-dioxo-4,5-dihydro-4lambda~6~-benzo[c]thieno[3,4-e][1,2]thiazine-1-
Supplier: Apollo Scientific
OR25277-1G OR25277-5G
APOSOR25277-1GEA 87 EUR
APOSOR25277-1G APOSOR25277-5G
Ethyl 5-methyl-3-(methylthio)-4,4-dioxo-4,5-dihydro-4lambda~6~-benzo[c]thieno[3,4-e][1,2]thiazine-1-
Ethyl 5-methyl-3-(methylthio)-4,4-dioxo-4,5-dihydro-4lambda~6~-benzo[c]thieno[3,4-e][1,2]thiazine-1-
Formula: C₁₅H₁₅NO₄S₃
MW: 369.479 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00085073
CAS Number: 175276-42-3