2-(2-Hydroxyprop-2-yl)pyridine
Supplier: Apollo Scientific
Warning
Synonyms: 2-(Pyridin-2-yl)propan-2-ol
OR25562-250MG OR25562-1G
APOSOR25562-250MGEA 43.5 EUR
APOSOR25562-250MG APOSOR25562-1G
2-(2-Hydroxyprop-2-yl)pyridine
2-(2-Hydroxyprop-2-yl)pyridine
Formula: C₈H₁₁NO
MW: 137.179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00209259
CAS Number: 37988-38-8