3-(Methylthio)-1-phenyl-1H-pyrazole-4-carbonitrile
Supplier: Apollo Scientific
OR25578-1G OR25578-5G
APOSOR25578-1GEA 102 EUR
APOSOR25578-1G APOSOR25578-5G
3-(Methylthio)-1-phenyl-1H-pyrazole-4-carbonitrile
3-(Methylthio)-1-phenyl-1H-pyrazole-4-carbonitrile
Formula: C₁₁H₉N₃S
MW: 215.274 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00173847
CAS Number: 175203-46-0