2-Bromo-5-chlorobenzo[b]thiophene-3-carboxaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-5-chloro-3-formyl-1-benzothiophene
OR26058-1G OR26058-250MG
APOSOR26058-1GEA 224 EUR
APOSOR26058-1G APOSOR26058-250MG
2-Bromo-5-chlorobenzo[b]thiophene-3-carboxaldehyde
2-Bromo-5-chlorobenzo[b]thiophene-3-carboxaldehyde
Formula: C₉H₄BrClOS
MW: 275.549 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01570694
CAS Number: 680212-97-9