4-(dimethylamino)-1,1-dimethoxybut-3-en-2-one 98%
Supplier: Apollo Scientific
Synonyms: 4-(Dimethylamino)-1,1-dimethoxybut-3-en-2-one
OR27385-25G OR27385-5G
APOSOR27385-25GEA 221 EUR
APOSOR27385-25G APOSOR27385-5G
4-(dimethylamino)-1,1-dimethoxybut-3-en-2-one 98%
4-(dimethylamino)-1,1-dimethoxybut-3-en-2-one
Formula: C₈H₁₅NO₃
MW: 173.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00085024
CAS Number: 67751-23-9