2,6-Dimethyl-3-hydroxypyridine
Supplier: Apollo Scientific
Warning
Synonyms: 2,6-Dimethylpyridin-3-ol, 3-Hydroxy-2,6-lutidine
OR27677-10G OR27677-1G OR27677-5G
APOSOR27677-10GEA 154 EUR
APOSOR27677-10G APOSOR27677-1G APOSOR27677-5G
2,6-Dimethyl-3-hydroxypyridine
2,6-Dimethyl-3-hydroxypyridine
Formula: C₇H₉NO
MW: 123.152 g/mol
Melting Pt: 210…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00228692
CAS Number: 1122-43-6