1-(2,5-Dichloro-4-nitrophenyl)-2,5-dimethyl-1H-pyrrole
Supplier: Apollo Scientific
OR27703-10G OR27703-1G OR27703-5G
APOSOR27703-10GEA 1250 EUR
APOSOR27703-10G APOSOR27703-1G APOSOR27703-5G
1-(2,5-Dichloro-4-nitrophenyl)-2,5-dimethyl-1H-pyrrole
1-(2,5-Dichloro-4-nitrophenyl)-2,5-dimethyl-1H-pyrrole
Formula: C₁₂H₁₀Cl₂N₂O₂
MW: 285.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01570806
CAS Number: 302901-02-6