trans-4-Chloro-4'-methylchalcone
Supplier: Apollo Scientific
Warning
Synonyms: (2E)-3-(4-Chlorophenyl)-1-(4-methylphenyl)prop-2-en-1-one, (2E)-3-(4-Chlorophenyl)-1-(4-methylphenyl)-1-oxoprop-2-ene
OR27882-1G OR27882-250MG
APOSOR27882-1GEA 140 EUR
APOSOR27882-1G APOSOR27882-250MG
trans-4-Chloro-4'-methylchalcone
trans-4-Chloro-4'-methylchalcone
Formula: C₁₆H₁₃ClO
MW: 256.727 g/mol
Melting Pt: 143…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00016346
CAS Number: 37620-37-4