4-(tert-Butyl)benzoyl isothiocyanate
Supplier: Apollo Scientific
Danger
Synonyms: 1-(tert-Butyl)-4-(isothiocyanatocarbonyl)benzene
OR28448-1G OR28448-5G
APOSOR28448-1GEA 248 EUR
APOSOR28448-1G APOSOR28448-5G
4-(tert-Butyl)benzoyl isothiocyanate
4-(tert-Butyl)benzoyl isothiocyanate
Formula: C₁₂H₁₃NOS
MW: 219.303 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00175795
CAS Number: 680214-97-5
UN: 2923
ADR: 8+6.1,III