4-[2-(Bromomethyl)-1,3-dioxolan-2-yl]benzonitrile
Supplier: Apollo Scientific
Warning
Synonyms: 2-(Bromomethyl)-2-(4-cyanophenyl)-1,3-dioxolane
OR29054-1G OR29054-5G OR29054-10G
APOSOR29054-1GEA 37.7 EUR
APOSOR29054-1G APOSOR29054-5G APOSOR29054-10G
4-[2-(Bromomethyl)-1,3-dioxolan-2-yl]benzonitrile
4-[2-(Bromomethyl)-1,3-dioxolan-2-yl]benzonitrile
Formula: C₁₁H₁₀BrNO₂
MW: 268.107 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00052887
CAS Number: 60207-22-9