α,2,4-Trichlorobenzaldoxime
Supplier: Apollo Scientific
Danger
OR29855-1G OR29855-5G OR29855-250MG
APOSOR29855-1GEA 82.8 EUR
APOSOR29855-1G APOSOR29855-5G APOSOR29855-250MG
α,2,4-Trichlorobenzaldoxime
α,2,4-Trichlorobenzaldoxime
Formula: C₇H₄Cl₃NO
Melting Pt: 93…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00192643
CAS Number: 29203-60-9
UN: 1759
ADR: 8,III