5-Bromo-3-methylisoquinoline
Supplier: Apollo Scientific
Warning
OR300085-1G OR300085-250MG OR300085-500MG
APOSOR300085-1GEA 894 EUR
APOSOR300085-1G APOSOR300085-250MG APOSOR300085-500MG
5-Bromo-3-methylisoquinoline
5-Bromo-3-methylisoquinoline
Formula: C₁₀H₈BrN
MW: 222.081 g/mol
Melting Pt: 40…42 °C
Storage Temperature: Ambient
MDL Number: MFCD18260472
CAS Number: 16552-67-3