Ethyl 1-methyl-5-propyl-1H-pyrazole-3-carboxylate
Supplier: Apollo Scientific
Warning
OR300143-1G OR300143-250MG
APOSOR300143-1GEA 244 EUR
APOSOR300143-1G APOSOR300143-250MG
Ethyl 1-methyl-5-propyl-1H-pyrazole-3-carboxylate
Ethyl 1-methyl-5-propyl-1H-pyrazole-3-carboxylate
Formula: C₁₀H₁₆N₂O₂
MW: 196.246 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115092
CAS Number: 247583-69-3