6-Chloro-1,7-naphthyridin-4-ol 95%
Supplier: Apollo Scientific
Warning
OR300199-250MG OR300199-1G
APOSOR300199-250MGEA 531 EUR
APOSOR300199-250MG APOSOR300199-1G
6-Chloro-1,7-naphthyridin-4-ol 95%
6-Chloro-1,7-naphthyridin-4-ol
Formula: C₈H₅ClN₂O
MW: 180.591 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28129182
CAS Number: 1214241-98-1