1-(3-Bromo-5-methylphenyl)ethanamine hydrochloride
Supplier: Apollo Scientific
Warning
Synonyms: 1-(3-Bromo-5-methylphenyl)ethan-1-amine hydrochloride
OR300232-250MG OR300232-100MG
APOSOR300232-250MGEA 364 EUR
APOSOR300232-250MG APOSOR300232-100MG
1-(3-Bromo-5-methylphenyl)ethanamine hydrochloride
1-(3-Bromo-5-methylphenyl)ethanamine hydrochloride
Formula: C₉H₁₂BrN·ClH
MW: 250.563 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31561266
CAS Number: 2227272-47-9