1-(Quinolin-8-yl)ethanone
Supplier: Apollo Scientific
|
Warning
|
OR300272-1G
OR300272-250MG
OR300272-5G
APOSOR300272-1GEA
284
EUR
APOSOR300272-1G
APOSOR300272-250MG
APOSOR300272-5G
1-(Quinolin-8-yl)ethanone
1-(Quinolin-8-yl)ethanone
Formula:
C₁₁H₉NO MW: 171.195 g/mol Boiling Pt: 176 °C (12 mm Hg) Melting Pt: 42…45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047620 CAS Number: 56234-20-9 |