4,5,6,7-Tetrahydro-5-methylthiazolo[5,4-c]pyridin-2-amine
Supplier: Apollo Scientific
Warning
Synonyms: 5-Methyl-4,5,6,7-tetrahydrothiazolo[5,4-c]pyridin-2-amine
OR300274-1G OR300274-250MG
APOSOR300274-1GEA 238 EUR
APOSOR300274-1G APOSOR300274-250MG
4,5,6,7-Tetrahydro-5-methylthiazolo[5,4-c]pyridin-2-amine
4,5,6,7-Tetrahydro-5-methylthiazolo[5,4-c]pyridin-2-amine
Formula: C₇H₁₁N₃S
MW: 169.247 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02854749
CAS Number: 17899-48-8