5-Methyl-4,5,6,7-tetrahydrothiazolo[4,5-c]pyridine-2-carboxylic acid hydrochloride 95%
Supplier: Apollo Scientific
OR300279-1G OR300279-500MG
APOSOR300279-1GEA 868 EUR
APOSOR300279-1G APOSOR300279-500MG
5-Methyl-4,5,6,7-tetrahydrothiazolo[4,5-c]pyridine-2-carboxylic acid hydrochloride 95%
5-Methyl-4,5,6,7-tetrahydrothiazolo[4,5-c]pyridine-2-carboxylic acid hydrochloride
Formula: C₈H₁₀N₂O₂S·ClH
MW: 234.703 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26680605
CAS Number: 693771-79-8