7,8-Dihydro-7,7-dimethyl-5H-pyrido[3,2-b]indol-9(6H)-one
Supplier: Apollo Scientific
Warning
OR300312-100MG OR300312-250MG
APOSOR300312-100MGEA 163 EUR
APOSOR300312-100MG APOSOR300312-250MG
7,8-Dihydro-7,7-dimethyl-5H-pyrido[3,2-b]indol-9(6H)-one
7,8-Dihydro-7,7-dimethyl-5H-pyrido[3,2-b]indol-9(6H)-one
Formula: C₁₃H₁₄N₂O
MW: 214.263 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30490125
CAS Number: 1595028-26-4