1,6-Naphthyridine-5-carbonitrile
Supplier: Apollo Scientific
Warning
OR300345-1G OR300345-250MG
APOSOR300345-1GEA 1080 EUR
APOSOR300345-1G APOSOR300345-250MG
1,6-Naphthyridine-5-carbonitrile
1,6-Naphthyridine-5-carbonitrile
Formula: C₉H₅N₃
MW: 155.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06762717
CAS Number: 28694-40-8