7-Chloro-3-iodo-1H-pyrazolo[4,3-c]pyridine
Supplier: Apollo Scientific
Warning
OR300377-1G OR300377-250MG
APOSOR300377-1GEA 455 EUR
APOSOR300377-1G APOSOR300377-250MG
7-Chloro-3-iodo-1H-pyrazolo[4,3-c]pyridine
7-Chloro-3-iodo-1H-pyrazolo[4,3-c]pyridine
Formula: C₆H₃ClIN₃
MW: 279.466 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22380448
CAS Number: 1357946-98-5