tert-Butyl 6-chloro-3-methylpyridin-2-ylcarbamate
Supplier: Apollo Scientific
Warning
Synonyms: tert-Butyl (6-chloro-3-methylpyridin-2-yl)carbamate
OR300402-1G OR300402-250MG
APOSOR300402-1GEA 328 EUR
APOSOR300402-1G APOSOR300402-250MG
tert-Butyl 6-chloro-3-methylpyridin-2-ylcarbamate
tert-Butyl 6-chloro-3-methylpyridin-2-ylcarbamate
Formula: C₁₁H₁₅ClN₂O₂
MW: 242.702 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16036499
CAS Number: 679392-20-2