3,6-Dibromo-1H-pyrazolo[4,3-c]pyridine
Supplier: Apollo Scientific
Danger
OR300406-1G OR300406-250MG
APOSOR300406-1GEA 364 EUR
APOSOR300406-1G APOSOR300406-250MG
3,6-Dibromo-1H-pyrazolo[4,3-c]pyridine
3,6-Dibromo-1H-pyrazolo[4,3-c]pyridine
Formula: C₆H₃Br₂N₃
MW: 276.916 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22380429
CAS Number: 1357946-34-9
UN: 2811
ADR: 6.1,III