5-[3-(tert-Butyl)-1-(3-methylbenzyl)-1H-pyrazol-5-yl]-1,3,4-oxadiazole-2-thiol
Supplier: Apollo Scientific
OR30052-10G OR30052-1G OR30052-5G
APOSOR30052-10GEA 691 EUR
APOSOR30052-10G APOSOR30052-1G APOSOR30052-5G
5-[3-(tert-Butyl)-1-(3-methylbenzyl)-1H-pyrazol-5-yl]-1,3,4-oxadiazole-2-thiol
5-[3-(tert-Butyl)-1-(3-methylbenzyl)-1H-pyrazol-5-yl]-1,3,4-oxadiazole-2-thiol
Formula: C₁₇H₂₀N₄OS
MW: 328.432 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01765438
CAS Number: 306937-16-6