3-tert-Butyl-4-cyano-1H-pyrazole
Supplier: Apollo Scientific
Warning
Synonyms: 3-tert-butyl-4-cyanopyrazole
OR300793-1G OR300793-50MG OR300793-100MG OR300793-250MG OR300793-500MG
APOSOR300793-1GEA 249 EUR
APOSOR300793-1G APOSOR300793-50MG APOSOR300793-100MG APOSOR300793-250MG APOSOR300793-500MG
3-tert-Butyl-4-cyano-1H-pyrazole
3-tert-Butyl-4-cyano-1H-pyrazole
Formula: C₈H₁₁N₃
MW: 149.193 g/mol
Melting Pt: 131…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00121111
CAS Number: 875554-79-3