Methyl 3-amino-2-bromobenzoate
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2-Bromo-3-(methoxycarbonyl)aniline
OR301136-1G
OR301136-5G
OR301136-10G
APOSOR301136-1GEA
26.1
EUR
APOSOR301136-1G
APOSOR301136-5G
APOSOR301136-10G
Methyl 3-amino-2-bromobenzoate
Methyl 3-amino-2-bromobenzoate
Formula:
C₈H₈BrNO₂ Density: 1.583 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD03844437 CAS Number: 106896-48-4 UN: 2810 ADR: 6.1,III |