2,3-Dihydrobenzofuran-6-ol, Technical Grade
Supplier: Apollo Scientific
Warning
OR301201-1G OR301201-250MG
APOSOR301201-1GEA 625 EUR
APOSOR301201-1G APOSOR301201-250MG
2,3-Dihydrobenzofuran-6-ol, Technical Grade
2,3-Dihydrobenzofuran-6-ol
Formula: C₈H₈O₂
MW: 136.148 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18378253
CAS Number: 23681-89-2