4-(4-Formyl-2-thiazolyl)-1-piperidinecarboxylic acid, 1,1-dimethylethyl ester 97%
Supplier: Apollo Scientific
Warning
OR301228-1G OR301228-250MG
APOSOR301228-1GEA 364 EUR
APOSOR301228-1G APOSOR301228-250MG
4-(4-Formyl-2-thiazolyl)-1-piperidinecarboxylic acid, 1,1-dimethylethyl ester 97%
4-(4-Formyl-2-thiazolyl)-1-piperidinecarboxylic acid, 1,1-dimethylethyl ester
Formula: C₁₄H₂₀N₂O₃S
Melting Pt: 91…93 °C
Storage Temperature: Ambient
MDL Number: MFCD06658981
CAS Number: 869901-02-0