7-Oxa-2-azaspiro[3.5]nonane hydrochloride 95+%
Supplier: Apollo Scientific
Warning
OR301500-1G OR301500-250MG
APOSOR301500-1GEA 203 EUR
APOSOR301500-1G APOSOR301500-250MG
7-Oxa-2-azaspiro[3.5]nonane hydrochloride 95+%
7-Oxa-2-azaspiro[3.5]nonane hydrochloride
Formula: C₇H₁₃NO·ClH
MW: 163.645 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22421803
CAS Number: 1417633-09-0