1-Methylcyclohexanecarbonyl chloride
Supplier: Apollo Scientific
OR302386-1G OR302386-250MG OR302386-500MG
APOSOR302386-1GEA 396 EUR
APOSOR302386-1G APOSOR302386-250MG APOSOR302386-500MG
1-Methylcyclohexanecarbonyl chloride
1-Methylcyclohexanecarbonyl chloride
Formula: C₈H₁₃ClO
Storage Temperature: Ambient
MDL Number: MFCD00019346
CAS Number: 2890-61-1