4-Bromo-1H-pyrrolo[3,2-c]pyridine 95%
Supplier: Apollo Scientific
OR302726-100MG OR302726-250MG
APOSOR302726-100MGEA 120 EUR
APOSOR302726-100MG APOSOR302726-250MG
4-Bromo-1H-pyrrolo[3,2-c]pyridine 95%
4-Bromo-1H-pyrrolo[3,2-c]pyridine
Formula: C₇H₅BrN₂
MW: 197.032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08690131
CAS Number: 1000342-68-6