5-Methyl-3-phenylisoxazol-4-amine
Supplier: Apollo Scientific
Warning
Synonyms: 5-Methyl-3-phenyl-1,2-oxazol-4-amine, 4-Amino-5-methyl-3-phenylisoxazole
OR30303-1G OR30303-250MG
APOSOR30303-1GEA 132 EUR
APOSOR30303-1G APOSOR30303-250MG
5-Methyl-3-phenylisoxazol-4-amine
5-Methyl-3-phenylisoxazol-4-amine
Formula: C₁₀H₁₀N₂O
MW: 174.199 g/mol
Boiling Pt: 131…133 °C (0.5 mm Hg)
Melting Pt: 51…53 °C
Storage Temperature: Ambient
MDL Number: MFCD02677726
CAS Number: 21169-65-3
UN: 2811
ADR: 6.1,III