2-(4-Nitrophenyl)-4H,5H,6H,7H-furo[3,2-c]pyridine
Supplier: Apollo Scientific
OR303154-1G OR303154-500MG
APOSOR303154-1GEA 336 EUR
APOSOR303154-1G APOSOR303154-500MG
2-(4-Nitrophenyl)-4H,5H,6H,7H-furo[3,2-c]pyridine
2-(4-Nitrophenyl)-4H,5H,6H,7H-furo[3,2-c]pyridine
Formula: C₁₃H₁₂N₂O₃
MW: 244.246 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22689166
CAS Number: 1192968-13-0