8-Hydroxy-1H,2H,3H,4H-cyclopenta[c]chromen-4-one
Supplier: Apollo Scientific
OR303182-1G OR303182-500MG
APOSOR303182-1GEA 180 EUR
APOSOR303182-1G APOSOR303182-500MG
8-Hydroxy-1H,2H,3H,4H-cyclopenta[c]chromen-4-one
8-Hydroxy-1H,2H,3H,4H-cyclopenta[c]chromen-4-one
Formula: C₁₂H₁₀O₃
MW: 202.206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08691761
CAS Number: 131526-88-0