2,2-Dimethyl-6-nitro-2H,3H,4H-pyrido[3,2-b][1,4]oxazin-3-one
Supplier: Apollo Scientific
Synonyms: 2,2-Dimethyl-6-nitro-2H-pyrido[3,2-b][1,4]oxazin-3(4H)-one
OR303183-1G OR303183-500MG
APOSOR303183-1GEA 180 EUR
APOSOR303183-1G APOSOR303183-500MG
2,2-Dimethyl-6-nitro-2H,3H,4H-pyrido[3,2-b][1,4]oxazin-3-one
2,2-Dimethyl-6-nitro-2H,3H,4H-pyrido[3,2-b][1,4]oxazin-3-one
Formula: C₉H₉N₃O₄
MW: 223.185 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10000811
CAS Number: 1002726-59-1