2-(Aminomethyl)-2-methyl-1,3-dioxolane
Supplier: Apollo Scientific
Warning
Synonyms: (2-Methyl-1,3-dioxolan-2-yl)methanamine, (2-Methyl-1,3-dioxolan-2-yl)methylamine
OR303185-1G OR303185-100MG OR303185-250MG
APOSOR303185-1GEA 183 EUR
APOSOR303185-1G APOSOR303185-100MG APOSOR303185-250MG
2-(Aminomethyl)-2-methyl-1,3-dioxolane
2-(Aminomethyl)-2-methyl-1,3-dioxolane
Formula: C₅H₁₁NO₂
MW: 117.146 g/mol
Boiling Pt: 73 °C (31 mm Hg)
Melting Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD10698199
CAS Number: 3289-19-8