3,5-Dibromophenoxytrimethylsilane
Supplier: Apollo Scientific
OR303200-1G
OR303200-500MG
APOSOR303200-1GEA
270
EUR
APOSOR303200-1G
APOSOR303200-500MG
3,5-Dibromophenoxytrimethylsilane
3,5-Dibromophenoxytrimethylsilane
Formula:
C₉H₁₂Br₂OSi MW: 324.084 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23136036 CAS Number: 856766-98-8 |