2-Oxo-5-(pyridin-4-yl)-1,2-dihydropyridine-3-carboxylic acid
Supplier: Apollo Scientific
Warning
OR303239-1G OR303239-500MG
APOSOR303239-1GEA 671 EUR
APOSOR303239-1G APOSOR303239-500MG
2-Oxo-5-(pyridin-4-yl)-1,2-dihydropyridine-3-carboxylic acid
2-Oxo-5-(pyridin-4-yl)-1,2-dihydropyridine-3-carboxylic acid
Formula: C₁₁H₈N₂O₃
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD23697510
CAS Number: 62749-61-5