5-Iodo-6-methoxy-1,2,3,4-tetrahydronaphthalen-1-one
Supplier: Apollo Scientific
OR303313-1G OR303313-500MG
APOSOR303313-1GEA 538 EUR
APOSOR303313-1G APOSOR303313-500MG
5-Iodo-6-methoxy-1,2,3,4-tetrahydronaphthalen-1-one
5-Iodo-6-methoxy-1,2,3,4-tetrahydronaphthalen-1-one
Formula: C₁₁H₁₁IO₂
MW: 302.108 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18648151
CAS Number: 436091-93-9