(3S,4S,5S,7S)-6-Methyl-2-oxa-6-azatricyclo[3.3.1.03,7]nonan-4-ol

Supplier: Apollo Scientific
Warning

OR303348-1G OR303348-500MG
APOSOR303348-1GEA 244 EUR
APOSOR303348-1G APOSOR303348-500MG
(3S,4S,5S,7S)-6-Methyl-2-oxa-6-azatricyclo[3.3.1.03,7]nonan-4-ol
(3S,4S,5S,7S)-6-Methyl-2-oxa-6-azatricyclo[3.3.1.03,7]nonan-4-ol
Formula: C₈H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD07783424
CAS Number: 487-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR