2-Bromo-3-nonylthiophene

Supplier: Apollo Scientific

OR303356-10G OR303356-1G OR303356-5G
APOSOR303356-10GEA 346 EUR
APOSOR303356-10G APOSOR303356-1G APOSOR303356-5G
2-Bromo-3-nonylthiophene
2-Bromo-3-nonylthiophene
Formula: C₁₃H₂₁BrS
MW: 289.275 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20922797
CAS Number: 1398044-31-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR