2-Methyl-2,7-diazaspiro[4.4]nonan-1-one hydrochloride
Supplier: Apollo Scientific
OR304413-1G OR304413-250MG OR304413-500MG
APOSOR304413-1GEA 1560 EUR
APOSOR304413-1G APOSOR304413-250MG APOSOR304413-500MG
2-Methyl-2,7-diazaspiro[4.4]nonan-1-one hydrochloride
2-Methyl-2,7-diazaspiro[4.4]nonan-1-one hydrochloride
Formula: C₈H₁₄N₂O·ClH
MW: 190.671 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10700080
CAS Number: 1203686-07-0