2-Azaspiro[4.5]decane hydrochloride
Supplier: Apollo Scientific
OR304424-1G OR304424-5G
APOSOR304424-1GEA 745 EUR
APOSOR304424-1G APOSOR304424-5G
2-Azaspiro[4.5]decane hydrochloride
2-Azaspiro[4.5]decane hydrochloride
Formula: C₉H₁₇N·ClH
MW: 175.699 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05861558
CAS Number: 36392-74-2