(S)-5-Methyl-5-azaspiro[2.4]heptane-6-carboxylic acid
Supplier: Apollo Scientific
OR304438-1G OR304438-500MG
APOSOR304438-1GEA 1150 EUR
APOSOR304438-1G APOSOR304438-500MG
(S)-5-Methyl-5-azaspiro[2.4]heptane-6-carboxylic acid
(S)-5-Methyl-5-azaspiro[2.4]heptane-6-carboxylic acid
Formula: C₈H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD30146460
CAS Number: 1860033-47-1