5-(Bromomethyl)-1H-indazole, N1-BOC protected
Supplier: Apollo Scientific
Warning
Synonyms: tert-Butyl 5-(bromomethyl)-1H-indazole-1-carboxylate, 5-(Bromomethyl)-1-(tert-butoxycarbonyl)-1H-indazole
OR305067-250MG OR305067-500MG
APOSOR305067-250MGEA 944 EUR
APOSOR305067-250MG APOSOR305067-500MG
5-(Bromomethyl)-1H-indazole, N1-BOC protected
5-(Bromomethyl)-1H-indazole, N1-BOC protected
Formula: C₁₃H₁₅BrN₂O₂
MW: 311.174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13183030
CAS Number: 209804-25-1